Thermo Scientific Chemicals

Sodium bromide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 039426.14
25 g, Each
Thermo Scientific Chemicals

Sodium bromide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 039426.14
25 g, Each
Quantity
Catalog number: 039426.14
also known as 039426-14
Price (USD)
Price: 346.00
Online price: 293.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7647-15-6
IUPAC Name
sodium bromide
Molecular Formula
BrNa
InChI Key
JHJLBTNAGRQEKS-UHFFFAOYSA-M
SMILES
[Na+].[Br-]
Total Metal Impurities
0.001% max.
Particle Size
-10 Mesh
Appearance (Color)
White
Form
Beads
Residual water
< 100 ppm

Description

Sodium bromide is used in the preparation of bromides and as a source of bromide ion. It also finds its applications in drilling fluid additives, and weighting agent. It is employed in , Oil gas production, hydraulic fracturing, and fracking. It is also used in photography, to make bromides and organic chemicals, as a therapeutic sedative and hypnotic, and formerly as a veterinary medicine. It also finds its applications to control algae, bacteria, and fungi in water systems (pasteurizing, canning, pulp & paper manufacturing, and ornamental ponds); and used to repel moths from clothing and fleas from pets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text