Zirconium(IV) tert-butoxide, 99% (metals basis), Thermo Scientific Chemicals
Zirconium(IV) tert-butoxide, 99% (metals basis), Thermo Scientific Chemicals
Zirconium(IV) tert-butoxide, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Zirconium(IV) tert-butoxide, 99% (metals basis), Thermo Scientific Chemicals
製品番号(カタログ番号) 039547.06
または、製品番号039547-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
仕様
定量パーセント注記(metals basis)
バイルシュタイン3681870
CAS2081-12-1
化学物質名または材質Zirconium(IV) tert-butoxide
Yellow
さらに表示
It finds it application as a reagent for the preparation of heterometallic tin(II) or lead(II) zirconium alkoxides. It is used as a catalyst in cyanation process. Zirconium tert-butoxide (ZTB) precursor is used to deposit thin films of Zirconia and other zirconium containing films by atomic layer deposition and chemical vapor deposition methods. Zirconium oxide thin films can also be grown at low temperatures, ranging from 150°C to 300°C, by the presence of moisture along with ZTB or by UV-enhanced atomic layer deposition (UV-ALD) process.