Zinc meso-tetraphenylporphine
Zinc meso-tetraphenylporphine
Zinc meso-tetraphenylporphine
Thermo Scientific Chemicals

Zinc meso-tetraphenylporphine

CAS: 14074-80-7 | C44H28N4Zn | 678.12 g/mol
製品番号(カタログ番号) 039552.04
または、製品番号039552-04
価格(JPY)
-
数量:
2 g
一括またはカスタム形式をリクエストする
仕様
CAS14074-80-7
化学物質名または材質Zinc meso-tetraphenylporphine
Purple
式量678.1
IUPAC名5,10,15,20-tetraphenylporphyrin-22,24-diide;zinc
さらに表示
A porphyrin used in light emitting diodes. It has valuable biological applications in molecular biology, fluoroimmunoassay and new pharmaceutical developments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only