Zinc 2-ethylhexanoate, Zn ≈ 20%, cont. 1% diethylene glycolmonomethyl ether
Zinc 2-ethylhexanoate, Zn ≈ 20%, cont. 1% diethylene glycolmonomethyl ether
Zinc 2-ethylhexanoate, Zn ≈ 20%, cont. 1% diethylene glycolmonomethyl ether
Zinc 2-ethylhexanoate, Zn ≈ 20%, cont. 1% diethylene glycolmonomethyl ether
Thermo Scientific Chemicals

Zinc 2-ethylhexanoate, Zn ≈ 20%, cont. 1% diethylene glycolmonomethyl ether

CAS: 136-53-8 | C16H30O4Zn | 351.792 g/mol
製品番号(カタログ番号) 039560.36
価格(JPY)
-
見積もりを依頼する
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS136-53-8
IUPAC Namezinc(2+) bis(2-ethylhexanoate)
Molecular FormulaC16H30O4Zn
InChI KeyIFNXAMCERSVZCV-UHFFFAOYNA-L
SMILES[Zn++].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
さらに表示
Appearance (Color)Colorless to yellow
FormLiquid, viscous
Assay (Titration)17 to 23 wt% Zn
Zinc 2-ethylhexanoate is widely used in polyvinylchloride heat stabilizer, polyurethane catalyst and lubricant additive. It is an auxiliary drier used as pigments dispersing agent and prevents the paint film from webbing and frosting effects. It is involved in the preparation of highly porous dentritic zinc oxide nanostructures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only