Ytterbium(III) isopropoxide
Ytterbium(III) isopropoxide
Ytterbium(III) isopropoxide
Ytterbium(III) isopropoxide
Thermo Scientific Chemicals

Ytterbium(III) isopropoxide

CAS: 6742-69-4 | C9H24O3Yb | 353.342 g/mol
製品番号(カタログ番号) 039579.03
または、製品番号039579-03
価格(JPY)
-
見積もりを依頼する
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6742-69-4
IUPAC Nametris(propan-2-ol) ytterbium
Molecular FormulaC9H24O3Yb
InChI KeyIVGMSSQGEVYWID-UHFFFAOYSA-N
SMILES[Yb].CC(C)O.CC(C)O.CC(C)O
さらに表示
FormPowder
Appearance (Color)Pale cream
Total Metal Impurities≤1.0%
Yttrium(III) tris(isopropoxide) is a catalyst for stereoselective conjugate addition reactions, ring-opening polymerization, generation of reactive enolates, allylic substitution and cyano-phosphorylation of enones and synthesis of nanocomposites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only