Sodium stearate
Sodium stearate
Sodium stearate
Thermo Scientific Chemicals

Sodium stearate

CAS: 822-16-2 | C18H35NaO2 | 306.466 g/mol
製品番号(カタログ番号) 039648.A1
または、製品番号039648-A1
価格(JPY)
-
数量:
1 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS822-16-2
IUPAC Namesodium octadecanoate
Molecular FormulaC18H35NaO2
InChI KeyRYYKJJJTJZKILX-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCCCCCCC([O-])=O
さらに表示
FormPowder
CommentFree Fatty Acid (FFA): ≤3.0%
CommentMelting Point: ≥200°C
Loss on Drying≤3.0%
Appearance (Color)White
Used in adhesives and sealants, laundry and dishwashing products, plastic and rubber products. Used as surface active agents. It is the gelling agent for deodrant sticks. Used as waterproofing additives and ointments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only