Molybdenum powder, -325 mesh, Puratronic™, 99.997% (metals basis excluding W), W <130ppm, Thermo Scientific Chemicals
Molybdenum powder, -325 mesh, Puratronic™, 99.997% (metals basis excluding W), W <130ppm, Thermo Scientific Chemicals
Molybdenum powder, -325 mesh, Puratronic™, 99.997% (metals basis excluding W), W <130ppm, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molybdenum powder, -325 mesh, Puratronic™, 99.997% (metals basis excluding W), W <130ppm, Thermo Scientific Chemicals

製品番号(カタログ番号) 039686.22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7439-98-7
IUPAC Namemolybdenum
Molecular FormulaMo
InChI KeyZOKXTWBITQBERF-UHFFFAOYSA-N
SMILES[Mo]
さらに表示
Particle Size-325 mesh (Less than approximately 45 microns)
Trace MetalTotal metal impurities less than 0.003% by weight
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only