Molybdenum powder, APS typically 2-4 micron, 99.9% (metals basis), Thermo Scientific Chemicals
Molybdenum powder, APS typically 2-4 micron, 99.9% (metals basis), Thermo Scientific Chemicals
Molybdenum powder, APS typically 2-4 micron, 99.9% (metals basis), Thermo Scientific Chemicals
Molybdenum powder, APS typically 2-4 micron, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molybdenum powder, APS typically 2-4 micron, 99.9% (metals basis), Thermo Scientific Chemicals

From Hydrogen reduced oxide
製品番号(カタログ番号) 039687.22
価格(JPY)
-
見積もりを依頼する
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7439-98-7
IUPAC Namemolybdenum
Molecular FormulaMo
InChI KeyZOKXTWBITQBERF-UHFFFAOYSA-N
SMILES[Mo]
さらに表示
Appearance (Color)Gray to black
FormPowder
Total Metal Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only