Ethylenediaminetetraacetic acid disodium salt, 0.115N (0.0575M) Standardized solution
Ethylenediaminetetraacetic acid disodium salt, 0.115N (0.0575M) Standardized solution
Ethylenediaminetetraacetic acid disodium salt, 0.115N (0.0575M) Standardized solution
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid disodium salt, 0.115N (0.0575M) Standardized solution

CAS: 139-33-3 | C10H14N2Na2O8 | 336.21 g/mol
製品番号(カタログ番号) 039782.K2
または、製品番号039782-K2
価格(JPY)
-
数量:
1 L
一括またはカスタム形式をリクエストする
化学物質識別子
CAS139-33-3
IUPAC Namedisodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate
Molecular FormulaC10H14N2Na2O8
InChI KeyZGTMUACCHSMWAC-UHFFFAOYSA-L
SMILES[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
さらに表示
FormLiquid
Assay from Supplier's CofA0.11477 N - 0.11523 N
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only