Thermo Scientific Chemicals
USA Standard Sieve, 200 Mesh
Catalog number: 039991.NR
5.08cm, 20.3 cm, Each
Thermo Scientific Chemicals
USA Standard Sieve, 200 Mesh
Catalog number: 039991.NR
5.08cm, 20.3 cm, Each
Depth (Metric)
Diameter (Metric) Outer
Catalog number: 039991.NR
also known as 039991-NR
Price (USD)
Quantity
-
Form
USA Standard Sieve (200 Mesh).
Diameter
20cm
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Brass frame, stainless steel mesh sieves meet ASTM E11 requirements and all current amendments
  • Sieves are full height
Sieve Depths
  • 3 in. diameter sieves are 1 in. deep
  • 8 in. diameter sieves are 2 in. deep
  • 12 in.diameter sieves are 3 in. deep