Thermo Scientific Chemicals

Potassium hexafluorophosphate, 99% min, Thermo Scientific Chemicals

Catalog number: 040030.22
100 g, Each
Thermo Scientific Chemicals

Potassium hexafluorophosphate, 99% min, Thermo Scientific Chemicals

Catalog number: 040030.22
100 g, Each
Quantity
Catalog number: 040030.22
also known as 040030-22
Price (USD)
Price: 74.30
Online price: 63.65
Your price:
Quantity
-

Chemical Identifiers

CAS
17084-13-8
IUPAC Name
potassium hexafluoro-λ⁵-phosphanuide
Molecular Formula
F6KP
InChI Key
YZDGRYDIGCWVND-UHFFFAOYSA-N
SMILES
[K+].F[P-](F)(F)(F)(F)F
Form
Powder or crystals or crystalline powder
Assay from Supplier's CofA
PF₆ anion: ≥77.6% (theoretical 78.8%)
Appearance (Color)
White
Assay from Supplier's CofA
Free fluoride: ≤0.5%
Water Content
≤1.0%

Description

Potassium hexafluorophosphate is used to prepare 1-butyl-3-methylimidazolium hexaflurorphospahte and other organic hexafluorophospahte compounds such as alkylaryliodonium hexafluorophopahtes and triarylsulfonium hexafluorophosphates. It is also used in the cationic polymerization reactions as photo initiators. It involves in producing catalyst for peptide coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text