Thermo Scientific Chemicals

Copper(I) thiocyanate, 96% min, Thermo Scientific Chemicals

Catalog number: 040220.18
50 g, Each
Thermo Scientific Chemicals

Copper(I) thiocyanate, 96% min, Thermo Scientific Chemicals

Catalog number: 040220.18
50 g, Each
Quantity
Catalog number: 040220.18
also known as 040220-18
Price (USD)
Price: 42.80
Online price: 36.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1111-67-7
IUPAC Name
λ¹-copper(1+) cyanosulfanide
Molecular Formula
CCuNS
InChI Key
PDZKZMQQDCHTNF-UHFFFAOYSA-M
SMILES
[Cu+].[S-]C#N
Assay (unspecified)
96% min

Description

It is used as flame retardant. It is a good inorganic pigment, and is used as the antifouling paint for protecting the underwater surfaces of ships against vegetation. Manufacturing industrial chemicals, pharmaceuticals and also used in freezing solutions, electroplating , steel picking, printing, and corrosion inhibitor against acid gases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text