Thermo Scientific Chemicals

Calcium hydrogen phosphate, anhydrous, 98% min, Thermo Scientific Chemicals

Catalog number: 040232.A7
5 kg, Each
Thermo Scientific Chemicals

Calcium hydrogen phosphate, anhydrous, 98% min, Thermo Scientific Chemicals

Catalog number: 040232.A7
5 kg, Each
Quantity
Catalog number: 040232.A7
Price (USD)
Price: 344.00
Online price: 292.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7757-93-9
IUPAC Name
calcium hydrogen phosphate
Molecular Formula
CaHO4P
InChI Key
FUFJGUQYACFECW-UHFFFAOYSA-L
SMILES
[Ca++].OP([O-])([O-])=O
Assay (unspecified)
97.5% min

Description

It is used as a feed additive for livestock and poultry. It can promote feed digestion, put on animal’s weight, raise laying rate or producing milk rate, cure diseases such as rickets, osteomalacia and anemia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text