Thermo Scientific Chemicals

Antimony(III) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 040371.30
250 g, Each
Thermo Scientific Chemicals

Antimony(III) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 040371.30
250 g, Each
Quantity
Catalog number: 040371.30
also known as 040371-30
Price (USD)
Quantity
-

Chemical Identifiers

CAS
1345-04-6
IUPAC Name
diantimony(3+) trisulfanediide
Molecular Formula
S3Sb2
InChI Key
NVWBARWTDVQPJD-UHFFFAOYSA-N
SMILES
[S--].[S--].[S--].[Sb+3].[Sb+3]
Total Metal Impurities
0.1% max.

Description

Antimony Trisulfide is used in the production of pyrotechnics and as a yellow pigment for glass and porcelain. In Vacuum deposition. Used in preparation of Vermilion or yellow pigment, for antimony salts, percussion caps, camouflage paints, and in ruby glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text