Potassium hexachlororuthenate(IV), Premion™, 99.95% (metal basis), Ru 25.3% min
Potassium hexachlororuthenate(IV), Premion™, 99.95% (metal basis), Ru 25.3% min
Potassium hexachlororuthenate(IV), Premion™, 99.95% (metal basis), Ru 25.3% min
Thermo Scientific Chemicals

Potassium hexachlororuthenate(IV), Premion™, 99.95% (metal basis), Ru 25.3% min

CAS: 23013-82-3 | K2RuCl6
製品番号(カタログ番号) 040399.03
または、製品番号040399-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS23013-82-3
IUPAC Nameλ⁴-ruthenium(4+) dipotassium hexachloride
Molecular FormulaCl6K2Ru
InChI KeyXOLNQIIEFUNTQC-UHFFFAOYSA-H
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[K+].[K+].[Ru+4]
さらに表示
Total Metal Impurities0.05% max.
Assay (unspecified)Ru 25.3% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only