Thermo Scientific Chemicals

Platinum(IV) oxide, anhydrous, Premion™, 99.95% (metals basis), Pt 84.4% min, Thermo Scientific Chemicals

Catalog number: 040402.06
5 g, Each
Thermo Scientific Chemicals

Platinum(IV) oxide, anhydrous, Premion™, 99.95% (metals basis), Pt 84.4% min, Thermo Scientific Chemicals

Catalog number: 040402.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 040402.06
also known as 040402-06
Price (USD)
Price: 1,321.00
Online price: 1,122.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1314-15-4
IUPAC Name
dioxoplatinum
Molecular Formula
O2Pt
InChI Key
YKIOKAURTKXMSB-UHFFFAOYSA-N
SMILES
O=[Pt]=O
Form
Powder
Assay (special)
Pt 84.4-85.6%
Appearance (Color)
Brown
Total Metal Impurities
≤0.05%

Description

Platinum(IV) oxide is used as catalyst for hydrogenation reactions. It is applied in the hydrogenation of alkenes to alkanes, nitro compounds to amines and ketones to alcohols. It is also involved in the hydrogenolysis of cyclopropyl group in to an isopropyl group and in selective oxidation of primary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text