Iron foil, 0.1mm (0.004in) thick, 99.99% (metals basis)
Iron foil, 0.1mm (0.004in) thick, 99.99% (metals basis)
Iron foil, 0.1mm (0.004in) thick, 99.99% (metals basis)
Iron foil, 0.1mm (0.004in) thick, 99.99% (metals basis)
Thermo Scientific Chemicals

Iron foil, 0.1mm (0.004in) thick, 99.99% (metals basis)

CAS: 7439-89-6 | Fe | 55.85 g/mol
数量:
25 x 25 mm
50 x 50 mm
100 x 100 mm
製品番号(カタログ番号) 040493.GH
価格(JPY)
-
見積もりを依頼する
数量:
100 x 100 mm
一括またはカスタム形式をリクエストする
仕様
外見Gray foil
定量パーセント範囲99.99% (Metals basis)
CAS7439-89-6
ChEBICHEBI:82664
化学物質名または材質Iron
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only