Bis(cyclopentadienyl)tungsten dichloride, 99%
Thermo Scientific Chemicals

Bis(cyclopentadienyl)tungsten dichloride, 99%

CAS: 12184-26-8
製品番号(カタログ番号) 040521.03
または、製品番号040521-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS12184-26-8
Elemental AnalysisC: 30.89-31.51% (Theory: 31.20%)
Elemental AnalysisH: 2.59-2.65% (Theory: 2.62%)
Bis(cyclopentadienyl)tungsten dichloride a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only