Gadolinium(III) perchlorate hexahydrate, 50% w/w aq. soln., Reagent Grade
Gadolinium(III) perchlorate hexahydrate, 50% w/w aq. soln., Reagent Grade
Gadolinium(III) perchlorate hexahydrate, 50% w/w aq. soln., Reagent Grade
Thermo Scientific Chemicals

Gadolinium(III) perchlorate hexahydrate, 50% w/w aq. soln., Reagent Grade

CAS: 14017-52-8
製品番号(カタログ番号) 040571.NC
または、製品番号040571-NC
価格(JPY)
-
数量:
10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS14017-52-8
IUPAC Namegadolinium(3+) hydrate triperchlorate
Molecular FormulaCl3GdH2O13
InChI KeyTXABYLVQTHIELE-UHFFFAOYSA-K
SMILESO.[Gd+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
さらに表示
Concentration (weight/weight)50% min
[Gd(DO3A-Pr-ATP)(H2O)2] was synthesized by the reaction of DO3A-Pr-ATP with gadolinium(iii) perchlorate hydrate in water. Gadolinium(III) chelates approved by FDA for human imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only