Gold Thinfoil, 0.005mm (0.0002in) thick, 99.9% (metals basis)
Gold Thinfoil, 0.005mm (0.0002in) thick, 99.9% (metals basis)
Gold Thinfoil, 0.005mm (0.0002in) thick, 99.9% (metals basis)
Thermo Scientific Chemicals

Gold Thinfoil, 0.005mm (0.0002in) thick, 99.9% (metals basis)

CAS: 7440-57-5 | Au | 196.97 g/mol
数量:
25 x 25 mm
50 x 50 mm
製品番号(カタログ番号) 040739.FF
または、製品番号040739-FF
価格(JPY)
-
数量:
25 x 25 mm
一括またはカスタム形式をリクエストする
仕様
定量パーセント注記(metals basis)
CAS7440-57-5
ChEBICHEBI:30050
化学物質名または材質Gold Thinfoil
EINECS番号231-165-9
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only