Aluminum sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.999% (metals basis), Thermo Scientific Chemicals
Aluminum sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.999% (metals basis), Thermo Scientific Chemicals
Aluminum sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.999% (metals basis), Thermo Scientific Chemicals

Magnetron sputtering source
製品番号(カタログ番号)数量
040826.KS
または、製品番号040826-KS
1 Ea.
製品番号(カタログ番号) 040826.KS
または、製品番号040826-KS
価格(JPY)
-
数量:
1 Ea.
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
さらに表示
Assay from Supplier's CofA≥99.999% (metals basis)
Appearance (Color)Silver gray
FormSputtering target, 50.8mm (2.0in) diameter x 3.18mm (0.125in) thick.
Magnetron sputtering source

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only