Cobalt sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis)
Cobalt sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis)
Cobalt sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis)
Thermo Scientific Chemicals

Cobalt sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis)

CAS: 7440-48-4 | Co | 58.93 g/mol
製品番号(カタログ番号)数量
040835.KS1 Ea.
製品番号(カタログ番号) 040835.KS
価格(JPY)
-
数量:
1 Ea.
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-48-4
IUPAC Namecobalt
Molecular FormulaCo
InChI KeyGUTLYIVDDKVIGB-UHFFFAOYSA-N
SMILES[Co]
さらに表示
FormSputtering Target, 2.0 in. dia. x 0.25 in. thick
Total Metal Impurities≤0.05%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only