Nickel Chromium, sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)
Nickel Chromium, sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)
Nickel Chromium, sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)
Thermo Scientific Chemicals

Nickel Chromium, sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)

CAS: 11106-97-1 | CrNi | 110.69 g/mol
製品番号(カタログ番号)数量
040870.KS1 Ea.
製品番号(カタログ番号) 040870.KS
価格(JPY)
-
数量:
1 Ea.
一括またはカスタム形式をリクエストする
化学物質識別子
CAS11106-97-1
IUPAC Namechromium nickel
Molecular FormulaCrNi
InChI KeyVNNRSPGTAMTISX-UHFFFAOYSA-N
SMILES[Cr].[Ni]
さらに表示
Appearance (Color)Silver
Assay from Supplier's CofA≥99.95% (metals basis)
Assay from Supplier's CofANi:Cr; 80:20 wt%
FormSputtering target, 50.8mm (2.0in) diameter x 3.18mm (0.125in) thick.
Used as Magnetron sputtering source. The substrate bias during dc sputtering effectively reduces oxygen contamination in nichrome films. This in turn leads to an enhancement of the preferred orientation of the deposits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only