Thermo Scientific Chemicals
Stainless Steel wire, 0.51mm (0.02in) dia, Type 304, Thermo Scientific Chemicals
Catalog number: 040946.BZ
150 cm, Each
Thermo Scientific Chemicals
Stainless Steel wire, 0.51mm (0.02in) dia, Type 304, Thermo Scientific Chemicals
Catalog number: 040946.BZ
150 cm, Each
Quantity
Catalog number: 040946.BZ
also known as 040946-BZ
Price (USD)
Quantity
-
Description
It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov