Silicon(IV) nitride, MgO binder, sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.9% (metals basis)
Thermo Scientific Chemicals

Silicon(IV) nitride, MgO binder, sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.9% (metals basis)

 
製品番号(カタログ番号)数量
041088.KS1 Ea.
製品番号(カタログ番号) 041088.KS
価格(JPY)
-
数量:
1 Ea.
一括またはカスタム形式をリクエストする
仕様
定量パーセント注記(metals basis)
化学物質名または材質Silicon(IV) nitride, MgO binder, Sputtering Target
EINECS番号234-796-8
式量140.28
MDL番号MFCD00011230
さらに表示
Magnetron sputtering source

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only