Titanium nitride sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.5% (metals basis)
Titanium nitride sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.5% (metals basis)
Titanium nitride sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.5% (metals basis)
Thermo Scientific Chemicals

Titanium nitride sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.5% (metals basis)

CAS: 25583-20-4 | NTi | 61.87 g/mol
製品番号(カタログ番号)数量
041110.KS
または、製品番号041110-KS
1 Ea.
製品番号(カタログ番号) 041110.KS
または、製品番号041110-KS
価格(JPY)
-
数量:
1 Ea.
一括またはカスタム形式をリクエストする
仕様
定量パーセント注記(metals basis)
CAS25583-20-4
化学物質名または材質Titanium nitride sputtering target
Gray
EINECS番号247-117-5
さらに表示
Magnetron sputtering source

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only