Product Image
Thermo Scientific Chemicals
USA Standard Sieve, 60 Mesh
Brass frame and Stainless-steel mesh sieves
直径(メートル法)外径:
7.62 cm
20.3 cm
30.4 cm
製品番号(カタログ番号) 041200.NK
または、製品番号041200-NK
価格(JPY)
-
直径(メートル法)外径:
30.4 cm
一括またはカスタム形式をリクエストする
仕様
認証/コンプライアンスASTM E11
奥行き(ヤードポンド法)3 in.
奥行き(メートル法)7.62 cm
概要60 Mesh
直径(ヤードポンド法)外径12 in.
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Brass frame, stainless steel mesh sieves meet ASTM E11 requirements and all current amendments
  • Sieves are full height
Sieve Depths
  • 3 in. diameter sieves are 1 in. deep
  • 8 in. diameter sieves are 2 in. deep
  • 12 in.diameter sieves are 3 in. deep
RUO – Research Use Only