Carbon conductive cement adhesive
Thermo Scientific Chemicals
Carbon conductive cement adhesive
製品番号(カタログ番号)数量
041212.15
または、製品番号041212-15
30 g
製品番号(カタログ番号) 041212.15
または、製品番号041212-15
価格(JPY)
-
数量:
30 g
一括またはカスタム形式をリクエストする
仕様
Black
数量30 g
Unit SizeEach
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
よくあるご質問(FAQ)
How can I clean tools or instruments that were accidentally contaminated with Carbon conductive cement adhesive (Cat. No. 041212.15, 041212-15)?
Xylene or toluene can easily clean tools or instruments that are accidentally contaminated with Carbon conductive cement adhesive.
Please note that xylene and toluene are hazardous solvents and should only be used in adherence with the instructions provided on the relevant Safety Data Sheet for the solvent(s).
How do I use Carbon conductive cement adhesive (041212.15, 041212-15)?
A thin layer of Carbon conductive cement adhesive should be used for powders or very light specimens. If powders are going to be mounted, allow a short drying period (of several minutes) before mounting, so that the powder does not sink into the wet cement. Some trial and error for exact drying time may be necessary.
For bulk specimens, a heavier layer should be painted on the specimen stub. Heavier specimens can be placed directly into the wet cement.
Drying is the physical process of the evaporation of the organic solvents in the product. After fully drying, only carbon (with acrylic binding agents) will remain. Depending upon the thickness of the layer, the drying time varies from a few seconds (very thin films) up to 30 minutes (for a heavy coating).
What is the intended application for Carbon conductive cement adhesive (Cat. No. 041212.15, 041212-15)?
The Carbon conductive cement adhesive is an electrically conductive cement for specimen mounting in SEM work.