Polyvinyl alcohol, 98-99% hydrolyzed, high molecular weight
Polyvinyl alcohol, 98-99% hydrolyzed, high molecular weight
Polyvinyl alcohol, 98-99% hydrolyzed, high molecular weight
Polyvinyl alcohol, 98-99% hydrolyzed, high molecular weight
Thermo Scientific Chemicals

Polyvinyl alcohol, 98-99% hydrolyzed, high molecular weight

CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 041243.22
also known as 041243-22
Price (USD)
46.30
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
46.30
Each
Chemical Identifiers
CAS9002-89-5
Molecular Formula(C2H4O)n
SMILESOC(-*)C-*
MDL NumberMFCD00081922
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)< 5.0% volatiles
FormGranular powder
Identification (FTIR)Conforms
Appearance (Color)Pale cream
Polyvinyl alcohol is used in food industries as a binding and coating agent. Its viscosity favors the application as coating agent to tablets and capsules. Mixed-matrix membrane prepared by dispersion of NaA zeolite particles is used to study the pervaporative separation of water-butanol mixtures. Polyvinyl alcohol /gelatin based biocompatible polymeric scaffolds have been developed for 3D cancer models.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Frequently asked questions (FAQs)

What is the average molecular weight of Polyvinyl alcohol, 98-99% hydrolyzed, high molecular weight (Cat. No. 041243.xx)?

Polyvinyl alcohol, 98-99% hydrolyzed, high molecular weight has a typical average molecular weight of 126,000 g/mol.