Strontium hydroxide octahydrate, 99% (metals basis)
Strontium hydroxide octahydrate, 99% (metals basis)
Strontium hydroxide octahydrate, 99% (metals basis)
Thermo Scientific Chemicals

Strontium hydroxide octahydrate, 99% (metals basis)

CAS: 1311-10-0 | H18O10Sr | 265.75 g/mol
製品番号(カタログ番号) 041351.22
または、製品番号041351-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1311-10-0
IUPAC Namestrontium(2+) octahydrate dihydroxide
Molecular FormulaH18O10Sr
InChI KeyUJPWWRPNIRRCPJ-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.O.[OH-].[OH-].[Sr++]
さらに表示
Total Metal Impurities1% max.
Strontium hydroxide octahydrate is used in the sugar industry for its purification, oil and paint industry to improve its drying properties and as an active ingredient in depilatory hair removal products. It is also used as a raw material for manufacturing of other products of strontium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only