Aluminum Thinfoil, 8.0 micron (0.0003in) thick, 99.0% (metals basis)
Aluminum Thinfoil, 8.0 micron (0.0003in) thick, 99.0% (metals basis)
Aluminum Thinfoil, 8.0 micron (0.0003in) thick, 99.0% (metals basis)
Aluminum Thinfoil, 8.0 micron (0.0003in) thick, 99.0% (metals basis)
Thermo Scientific Chemicals

Aluminum Thinfoil, 8.0 micron (0.0003in) thick, 99.0% (metals basis)

Form: approx. 0.054g/50x50mm, not certified pinhole free | CAS: 7429-90-5 | Al | 26.98 g/mol
数量:
50 x 50 mm
100 x 100 mm
製品番号(カタログ番号) 041358.GH
または、製品番号041358-GH
価格(JPY)
-
見積もりを依頼する
数量:
100 x 100 mm
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
さらに表示
Total Metal Impurities≤1.0%
FormFoil, 8.0 micron (0.0003 in.) thick, not certified pinhole free
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only