Silver foil, 0.28mm (0.011in) thick, annealed, 99.9% (metals basis)
Silver foil, 0.28mm (0.011in) thick, annealed, 99.9% (metals basis)
Silver foil, 0.28mm (0.011in) thick, annealed, 99.9% (metals basis)
Thermo Scientific Chemicals

Silver foil, 0.28mm (0.011in) thick, annealed, 99.9% (metals basis)

CAS: 7440-22-4 | Ag | 107.87 g/mol
数量:
50 x 50 mm
100 x 100 mm
200 x 200 mm
製品番号(カタログ番号) 041451.GW
または、製品番号041451-GW
価格(JPY)
-
数量:
200 x 200 mm
一括またはカスタム形式をリクエストする
仕様
定量パーセント注記(metals basis)
CAS7440-22-4
ChEBICHEBI:9141
化学物質名または材質Silver foil
EINECS番号231-131-3
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only