Ethyl glycolate, 95%
Ethyl glycolate, 95%
Ethyl glycolate, 95%
Ethyl glycolate, 95%
Thermo Scientific Chemicals

Ethyl glycolate, 95%

CAS: 623-50-7 | C4H8O3 | 104.11 g/mol
製品番号(カタログ番号) 041475.09
または、製品番号041475-09
価格(JPY)
-
見積もりを依頼する
数量:
10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS623-50-7
Appearance (Color)Clear colorless
FormLiquid
Assay from Supplier's CofA≥94.0%

Ethyl glycolate, is an important building block for the preparation of agrochemicals and medicinal compounds. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff. It is also used in synthetic chemistry, discovery process and R&D. It also can be used as solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only