Thermo Scientific Chemicals

Sodium DL-lactate, 60% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: 041529.K2
1 L, Each
Thermo Scientific Chemicals

Sodium DL-lactate, 60% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: 041529.K2
1 L, Each
Quantity
Catalog number: 041529.K2
also known as 041529-K2
Price (USD)
Price: 150.00
Online price: 127.65
Your price:
Quantity
-

Chemical Identifiers

CAS
72-17-3
IUPAC Name
sodium 2-hydroxypropanoate
Molecular Formula
C3H5NaO3
InChI Key
NGSFWBMYFKHRBD-UHFFFAOYNA-M
SMILES
[Na+].CC(O)C([O-])=O
Assay (unspecified)
58 to 62%

Description

Useful chiral synthon; building block for depsipeptides.Sodium DL-lactate, 60% w/w aq. Soln is used as a food additive, plasticizer for casein, preservative, acidity regulator, bulking agent, and in shampoo and liquid soaps. It is also used for the treatment for arrhythmias and sympathomimetics. It is useful chiral synthon and building block for depsipeptides. It is used as a corrosion inhibitor in alcohol antifreeze mixtures, flavoring agent, emulsifier, corrosion inhibitor and pH control agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text