Glassy carbon spherical powder, 630-1000 micron, type 2, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Glassy carbon spherical powder, 630-1000 micron, type 2, Thermo Scientific Chemicals

Used as such in purifying metals, in electrodes, electrical devices & steel. Glassy Carbon Spherical Powder has numerous applications due its fundamental softness, including in lubricant formulations and as an additive to structural materials and metallic alloys.
Have Questions?
Catalog NumberQuantity
041530.30
also known as 041530-30
250 g
Catalog number 041530.30
also known as 041530-30
Price (USD)
580.65
Special offer
Online exclusive
Ends: 28-Dec-2026
683.00
Save 102.35
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
580.65
Special offer
Online exclusive
Ends: 28-Dec-2026
683.00
Save 102.35
Each
Specifications
Chemical Name or MaterialGlassy carbon spherical powder
EINECS Number231-153-3
MDL NumberMFCD00133992
PackagingGlass bottle
Recommended StorageAmbient temperatures
View more

Glassy carbon spherical powder is used as such in purifying metals, in electrodes, electrical devices & steel. Glassy Carbon Spherical Powder has numerous applications due its fundamental softness, including in lubricant formulations and as an additive to structural materials and metallic alloys.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Frequently asked questions (FAQs)

What is the difference between type 1 glassy carbon and type 2 glassy carbon?

Type 1 is a "low temperature" glassy carbon produced by heat treatment at ~ 1,000-2,000°C and consists of discreet fragments of distorted graphene layers. In general, type 1 is less compressible, more resistant to oxidation, and less permeable to gasses. Type 2 is a"high temperature" glassy carbon produced by heat treatment at ~2,500-3,000°C and consists of broken or imperfect fullerene-like nanospheroids encased in a disordered, multilayered graphene matrix. Type 2 usually contains fewer impurities and has higher electrical and thermal conductivity.