Polyethylene glycol monomethylether, 350
Polyethylene glycol monomethylether, 350
Polyethylene glycol monomethylether, 350
Polyethylene glycol monomethylether, 350
Thermo Scientific Chemicals

Polyethylene glycol monomethylether, 350

CAS: 9004-74-4 | (C2H4O)nCH4O | 76.10 g/mol
製品番号(カタログ番号) 041560.36
または、製品番号041560-36
価格(JPY)
-
見積もりを依頼する
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS9004-74-4
Molecular Formula(C2H4O)nCH4O
InChI KeyXNWFRZJHXBZDAG-UHFFFAOYSA-N
SMILESCOCCO
MDL NumberMFCD00084416
Molecular Weight325 to 375 D
pH4.5 to 7.5 ( 5% aq sol at 25?C)
Residual water0.5% max
Polyethylene glycol monomethylether is used as enteric release coatings. It is also used for a series of polycarboxylate water reducing agent. It acts as a solvent for brake fluids. Further, it is used in the building materials industry and raw material for cement water reducing agent and strengthening agent. In addition to this, it is used in surfactants, polyester and polyurethane based paints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only