Thermo Scientific Chemicals

Sodium acetate hydrate, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals

Catalog number: 041685.22
100 g, Each
Thermo Scientific Chemicals

Sodium acetate hydrate, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals

Catalog number: 041685.22
100 g, Each
Quantity
Catalog number: 041685.22
also known as 041685-22
Price (USD)
Price: 682.00
Online price: 579.65
Your price:
Quantity
-

Chemical Identifiers

CAS
41484-91-7
IUPAC Name
sodium acetate
Molecular Formula
C2H3NaO2
InChI Key
VMHLLURERBWHNL-UHFFFAOYSA-M
SMILES
[Na+].CC([O-])=O
Total Metal Impurities
0.0015% max.

Description

Sodium acetate hydrate is used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text