Tungsten(VI) ethoxide
Tungsten(VI) ethoxide
Tungsten(VI) ethoxide
Thermo Scientific Chemicals
Tungsten(VI) ethoxide
Intermediate for sol-gel coatings | CAS: 62571-53-3 | C12H30O6W | 454.21 g/mol
製品番号(カタログ番号) 041795.06
または、製品番号041795-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
仕様
CAS62571-53-3
化学物質名または材質Tungsten(VI) ethoxide
White to Brown
DOT情報Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
式量454.21
さらに表示
Intermediate for sol-gel coatingsTungsten(VI) ethoxide is used in nano-materials, thin films, display and imaging, optics and glasses. It is also used for x-ray screen phosphors, fireproofing fabrics and in gas sensors. Also, it acts as an intermediate for sol-gel coatings.