Nickel tungsten oxide, 99.9% (metals basis excluding Co), Co <100ppm
Nickel tungsten oxide, 99.9% (metals basis excluding Co), Co <100ppm
Nickel tungsten oxide, 99.9% (metals basis excluding Co), Co <100ppm
Thermo Scientific Chemicals

Nickel tungsten oxide, 99.9% (metals basis excluding Co), Co <100ppm

CAS: 14177-51-6 | NiO4W | 306.529 g/mol
製品番号(カタログ番号) 041826.14
または、製品番号041826-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS14177-51-6
IUPAC Namenickel(2+) dioxotungstenbis(olate)
Molecular FormulaNiO4W
InChI KeyQLTKZXWDJGMCAR-UHFFFAOYSA-N
SMILES[Ni++].[O-][W]([O-])(=O)=O
さらに表示
Impurity contentCo <100ppm
Total Metal Impurities0.1% max.
Nickel tungsten oxide is used as photo catalysts, sensors and humidity dielectric resonators. It is considered as a new cathode material for asymmetric super capacitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only