1-Butanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles
1-Butanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles
1-Butanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles
Thermo Scientific Chemicals

1-Butanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles

CAS: 71-36-3 | C4H10O | 74.123 g/mol
数量:
250 mL
1 L
4 x 1 L
製品番号(カタログ番号) 041867.AK
または、製品番号041867-AK
価格(JPY)
-
数量:
250 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS71-36-3
IUPAC Namebutan-1-ol
Molecular FormulaC4H10O
InChI KeyLRHPLDYGYMQRHN-UHFFFAOYSA-N
SMILESCCCCO
さらに表示
FormLiquid, packaged under Argon in resealable ChemSeal? bottles.
Appearance (Color)Clear colorless, ≤10 APHA.
Assay (GC)GC-FID ≥99.9 % (corrected for water)
Residue on Evaporation≤ 10ppm
Insoluble materialPASS
さらに表示
1-Butanol is used as solvent for many organic reactions in pharmaceutical and chemical industry, as a solvent in coating industries and also used as a diluent/reactant in the manufacture of urea-formaldehyde and melamine-formaldehyde resins. It is used in the production of drugs, alkaloids, additive in floor cleaners, consumer products and stain removers. It is used as mobile phase in paper and thin layer chromatography. It is also used as an alternative for gasoline and diesel fuels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only