Sulfur in Crude Oil standard solution, Specpure™, 50,000μg/g (5.00%)
Thermo Scientific Chemicals

Sulfur in Crude Oil standard solution, Specpure™, 50,000μg/g (5.00%)

 
製品番号(カタログ番号)数量
041892.AE
または、製品番号041892-AE
100 mL
製品番号(カタログ番号) 041892.AE
または、製品番号041892-AE
価格(JPY)
-
数量:
100 mL
一括またはカスタム形式をリクエストする
仕様
化学物質名または材質Sulfur in Crude Oil standard solution
Black
DOT情報Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PETROLEUM CRUDE OIL
引火点25°C (77°F)
健康被害1H226-H335-H350
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only