Tricyclohexylphosphine, 98%
Tricyclohexylphosphine, 98%
Tricyclohexylphosphine, 98%
Thermo Scientific Chemicals

Tricyclohexylphosphine, 98%

CAS: 2622-14-2 | C18H33P | 280.436 g/mol
製品番号(カタログ番号) 041952.14
または、製品番号041952-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2622-14-2
IUPAC Nametricyclohexylphosphane
Molecular FormulaC18H33P
InChI KeyWLPUWLXVBWGYMZ-UHFFFAOYSA-N
SMILESC1CCC(CC1)P(C1CCCCC1)C1CCCCC1
さらに表示
Phosphorus-31 NMR> 97.5%
Used as an active pharmaceutical ingredient. Ligand used in the Pd-catalyzed coupling of malononitrile with aryl halides. Bulky phosphine ligand used with a Pd(0)-triolefinic macrocycle catalyst for Suzuki coupling of aryl bromides and chlorides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only