1,2-Dichloroethane-d{4}, 99% (Isotopic)
1,2-Dichloroethane-d{4}, 99% (Isotopic)
1,2-Dichloroethane-d{4}, 99% (Isotopic)
1,2-Dichloroethane-d{4}, 99% (Isotopic)
Thermo Scientific Chemicals

1,2-Dichloroethane-d{4}, 99% (Isotopic)

CAS: 17060-07-0 | C2H4Cl2 | 102.978 g/mol
数量:
1 g
5 g
5 x 1 g
製品番号(カタログ番号) 042281.06
または、製品番号042281-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS17060-07-0
IUPAC Name1,2-dichloroethane
Molecular FormulaC2H4Cl2
InChI KeyWSLDOOZREJYCGB-UHFFFAOYSA-N
SMILESClCCCl
さらに表示
FormLiquid
Assay from Supplier's CofA≥99.0% ¹H nmr
Water Content (Karl Fischer Titration)≤ 300ppm
Assay from Supplier's CofA≥99.0% Isotopic enrichment by ¹H nmr
Assay (GC)≥99.0%
さらに表示
1,2-Dichloroethane-d4 is widely used as a solvent for NMR analysis, and in the synthesis of poly(ethylene-d4 oxide) as a chain termination reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only