Molybdenum slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, 99.95% (metals basis)
Molybdenum slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, 99.95% (metals basis)
Molybdenum slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, 99.95% (metals basis)
Thermo Scientific Chemicals

Molybdenum slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, 99.95% (metals basis)

CAS: 7439-98-7 | Mo | 95.95 g/mol
製品番号(カタログ番号) 042360.18
または、製品番号042360-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
仕様
定量パーセント注記(metals basis)
CAS7439-98-7
ChEBICHEBI:28685
化学物質名または材質Molybdenum slug
EINECS番号231-107-2
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only