Buffer solution, pH 6.86 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable
Buffer solution, pH 6.86 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable
Buffer solution, pH 6.86 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable
Thermo Scientific Chemicals

Buffer solution, pH 6.86 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable

CAS: 1336-21-6 | H5NO | 35.05 g/mol
数量:
500 mL
1 L
4 L
製品番号(カタログ番号) 042419.K2
または、製品番号042419-K2
価格(JPY)
-
数量:
1 L
一括またはカスタム形式をリクエストする
仕様
CAS1336-21-6
ChEBICHEBI:18219
化学物質名または材質Buffer solution
Colorless
IUPAC名azanium;hydroxide
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only