Thermo Scientific Chemicals

Methoxytrimethylsilane, 97+%, Thermo Scientific Chemicals

Catalog number: 042464.18
50 g, Each
Thermo Scientific Chemicals

Methoxytrimethylsilane, 97+%, Thermo Scientific Chemicals

Catalog number: 042464.18
50 g, Each
Quantity
Catalog number: 042464.18
also known as 042464-18
Price (USD)
Price: 99.40
Online price: 84.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1825-61-2
IUPAC Name
methoxytrimethylsilane
Molecular Formula
C4H12OSi
InChI Key
POPACFLNWGUDSR-UHFFFAOYSA-N
SMILES
CO[Si](C)(C)C
Appearance (Color)
Clear colorless
Assay (GC)
≥97.0%
Refractive Index
1.3655-1.3705 @ 20°C
Form
Liquid

Description

Methoxytrimethylsilane plays an important role in the synthesis of organic compounds and used as water disgusting. It is used in the preparation of alfa-lithioalkoxysilanes by treating with tert-butyllithium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text