Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 117,000
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 117,000
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 117,000
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 117,000
Thermo Scientific Chemicals

Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 117,000

CAS: 9016-00-6 | (C2H6OSi)n | 74.15 g/mol
数量:
50 mL
250 mL
1 L
製品番号(カタログ番号) 042504.AK
または、製品番号042504-AK
価格(JPY)
-
見積もりを依頼する
数量:
250 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS9016-00-6
Molecular Formula(C2H6OSi)n
SMILESC[Si](C)(-*)O-*
MDL NumberMFCD00084411
Appearance (Color)Colorless
FormViscous liquid
Infrared spectrumConforms
Viscosity54000-66000 cSt
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only