Zirconium slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, 99.5% (metals basis excluding Hf), Hf nominal 3%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Zirconium slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, 99.5% (metals basis excluding Hf), Hf nominal 3%, Thermo Scientific Chemicals
Have Questions?
Quantity:
100 g
Catalog number 042508.22
also known as 042508-22
Price (USD)/ Each
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS7440-67-7
IUPAC Namezirconium
Molecular FormulaZr
InChI KeyQCWXUUIWCKQGHC-UHFFFAOYSA-N
SMILES[Zr]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery
FormSlug, 6.35mm (0.25in) diameter.
Assay from Supplier's CofA≥99.5% (metals basis excluding Hf), Hf nominal 3%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only