Sulfuric acid, 15% v/v aq. soln.
Sulfuric acid, 15% v/v aq. soln.
Sulfuric acid, 15% v/v aq. soln.
Thermo Scientific Chemicals

Sulfuric acid, 15% v/v aq. soln.

CAS: 7664-93-9 | H2O4S | 98.07 g/mol
製品番号(カタログ番号)数量
042554.K2
または、製品番号042554-K2
1 L
製品番号(カタログ番号) 042554.K2
または、製品番号042554-K2
価格(JPY)
-
数量:
1 L
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7664-93-9
IUPAC Namesulfuric acid
Molecular FormulaH2O4S
InChI KeyQAOWNCQODCNURD-UHFFFAOYSA-N
SMILESOS(O)(=O)=O
さらに表示
FormLiquid
Assay from Supplier's CofA15.00% v/v ? 0.75% v/v
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only