Germanium(IV) isopropoxide, 99.9% (metals basis)
Germanium(IV) isopropoxide, 99.9% (metals basis)
Germanium(IV) isopropoxide, 99.9% (metals basis)
Thermo Scientific Chemicals

Germanium(IV) isopropoxide, 99.9% (metals basis)

CAS: 21154-48-3 | C12H28GeO4 | 308.982 g/mol
製品番号(カタログ番号) 042673.14
または、製品番号042673-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
仕様
定量パーセント注記(metals basis)
CAS21154-48-3
化学物質名または材質Germanium (IV) isopropoxide
Colorless
密度1.025 g/mL
さらに表示
Germanium(IV) isopropoxide is used as precursor to prepare GeO2 organic silanes, which is used to study the application of optical wave guide. It is used in the synthesis of heterotactic polyactide and also involved in the macromolecule mediated synthesis of amorphous Germania.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only